MilliporeSigma
  • Regulation of mitogen-activated protein kinase activation by protein kinases A and C in a cell-free system.
Regulation of mitogen-activated protein kinase activation by protein kinases A and C in a cell-free system.
The Journal of biological chemistry (1994-10-07)
B VanRenterghem, M D Browning, J L Maller
ABSTRACT
Previously pp60v-src, cyclin A, p39mos, and maturation-promoting factor (composed of Cdc2 and cyclin B) have been shown to activate mitogen-activated protein kinase (MAPK) and MAPK kinase (MEK) in cell-free extracts of Xenopus oocytes. The pp60v-src pathway is dependent on a functional Ras signal whereas the cyclin/maturation-promoting factor pathway is not. Here we show that protein kinase C (PKC) is also able to stimulate MAPK in a Ras-dependent manner, but PKC is not necessary for signaling by pp60v-src. In addition, preincubation of extracts with cAMP-dependent protein kinase (PKA) blocks stimulation of MAPK by cyclin, p21V12ras, PKC, or pp60v-src, by at least 50%, but stimulation by c-Mos is unaffected. Furthermore, inhibition of endogenous PKA by the heat-stable PKA inhibitor is sufficient to stimulate MAPK activity in these extracts in the absence of protein synthesis and without dependence on a functional Ras protein. These results suggest that independent pp60v-src and PKC pathways converge at Ras and that PKA acts to block MAPK activation by both Ras-dependent and -independent signals.