MilliporeSigma
  • Activity of Alcohol Dehydrogenase and Aldehyde Dehydrogenase in Lung Cancer Cells.
Activity of Alcohol Dehydrogenase and Aldehyde Dehydrogenase in Lung Cancer Cells.
Anticancer research (2020-07-06)
Karolina Orywal, Wojciech Jelski, MirosŁaw Dariusz KozŁowski, Barbara Mroczko
ABSTRACT
The aim of this study was to define the alterations in the activity of alcohol dehydrogenase (ADH) isoenzymes and aldehyde dehydrogenase (ALDH) in normal and cancerous lung cells. Lung tissues were taken from 36 patients during surgical resection of cancer. The activities of tested enzymes were measured by spectrofluorometric method (ADH I, ADH II, total ALDH) and photometric method (ADH III, ADH IV, total ADH). The activities of class II and III ADH were significantly lower in lung cancer cells compared to histologically normal lung tissue. Reduced activity of isoenzyme class II ADH may affect disorders in retinoic acid biosynthesis, leading to its deficit. Lower ADH III activity may result in depletion of glutathione, and in initiation of oxidative stress, leading to cancer progression. These data suggest that alterations in ADH isoenzyme activities can contribute to carcinogenesis in human lungs.
Sigma-Aldrich
Fomepizole, 99%
Sigma-Aldrich
4-Methoxy-1-naphthaldehyde, 99%