MilliporeSigma
  • Proteomics and lipidomics analyses reveal modulation of lipid metabolism by perfluoroalkyl substances in liver of Atlantic cod (Gadus morhua).
Proteomics and lipidomics analyses reveal modulation of lipid metabolism by perfluoroalkyl substances in liver of Atlantic cod (Gadus morhua).
Aquatic toxicology (Amsterdam, Netherlands) (2020-09-06)
Karina Dale, Fekadu Yadetie, Mette Bjørge Müller, Daniela M Pampanin, Alejandra Gilabert, Xiaokang Zhang, Zhanna Tairova, Ane Haarr, Roger Lille-Langøy, Jan Ludvig Lyche, Cinta Porte, Odd André Karlsen, Anders Goksøyr
ABSTRACT
The aim of the present study was to investigate effects of defined mixtures of polycyclic aromatic hydrocarbons (PAHs) and perfluoroalkyl substances (PFASs), at low, environmentally relevant (1× = L), or high (20× = H) doses, on biological responses in Atlantic cod (Gadus morhua). To this end, farmed juvenile cod were exposed at day 0 and day 7 via intraperitoneal (i.p.) injections, in a two-week in vivo experiment. In total, there were 10 groups of fish (n = 21-22): two control groups, four separate exposure groups of PAH and PFAS mixtures (L, H), and four groups combining PAH and PFAS mixtures (L/L, H/L, L/H, H/H). Body burden analyses confirmed a dose-dependent accumulation of PFASs in cod liver and PAH metabolites in bile. The hepatosomatic index (HSI) was significantly reduced for three of the combined PAH/PFAS exposure groups (L-PAH/H-PFAS, H-PAH/L-PFAS, H-PAH/H-PFAS). Analysis of the hepatic proteome identified that pathways related to lipid degradation were significantly affected by PFAS exposure, including upregulation of enzymes in fatty acid degradation pathways, such as fatty acid β-oxidation. The increased abundances of enzymes in lipid catabolic pathways paralleled with decreasing levels of triacylglycerols (TGs) in the H-PFAS exposure group, suggest that PFAS increase lipid catabolism in Atlantic cod. Markers of oxidative stress, including catalase and glutathione S-transferase activities were also induced by PFAS exposure. Only minor and non-significant differences between exposure groups and control were found for cyp1a and acox1 gene expressions, vitellogenin concentrations in plasma, Cyp1a protein synthesis and DNA fragmentation. In summary, our combined proteomics and lipidomics analyses indicate that PFAS may disrupt lipid homeostasis in Atlantic cod.