MilliporeSigma
  • n-3 polyunsaturated fatty acids regulate chemerin in cultured adipocytes: role of GPR120 and derived lipid mediators.
n-3 polyunsaturated fatty acids regulate chemerin in cultured adipocytes: role of GPR120 and derived lipid mediators.
Food & function (2020-10-07)
N Sáinz, M Fernández-Galilea, A G V Costa, P L Prieto-Hontoria, G M Barraco, M J Moreno-Aliaga
ABSTRACT
Chemerin is a pro-inflammatory adipokine that is increased in obesity and associated with obesity-related comorbidities. The aim of this study was to investigate the effects of omega-3 polyunsaturated fatty acids, eicosapentaenoic and docosahexaenoic acids (EPA and DHA), on basal and tumor necrosis factor-α (TNF-α)-induced chemerin production in 3T3-L1 and human subcutaneous cultured adipocytes. The potential involvement of G protein-coupled receptor 120 (GPR120), as well as the actions of DHA-derived specialized proresolving lipid mediators (SPMs), resolvin D1 and D2 (RvD1 and RvD2) and maresin 1 (MaR1), were also evaluated. DHA significantly lowered both basal and TNF-α-stimulated chemerin production in 3T3-L1 and human adipocytes. EPA did not modify basal chemerin production, while it attenuated the induction of chemerin by TNF-α. Silencing of GPR120 using siRNA blocked the ability of DHA and EPA to reduce TNF-α-induced chemerin secretion. Interestingly, treatment with the DHA-derived SPMs RvD1, RvD2 and MaR1 also reversed the stimulatory effect of TNF-α on chemerin production in human adipocytes.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)