MilliporeSigma
  • Development of a Chimeric Antigen-Binding Fragment Directed Against Human Galectin-3 and Validation as an Immuno-Positron Emission Tomography Tracer for the Sensitive In Vivo Imaging of Thyroid Cancer.
Development of a Chimeric Antigen-Binding Fragment Directed Against Human Galectin-3 and Validation as an Immuno-Positron Emission Tomography Tracer for the Sensitive In Vivo Imaging of Thyroid Cancer.
Thyroid : official journal of the American Thyroid Association (2020-03-24)
Emanuel Peplau, Francesco De Rose, Sybille Reder, Markus Mittelhäuser, Giorgia Scafetta, Markus Schwaiger, Wolfgang A Weber, Armando Bartolazzi, Arne Skerra, Calogero D'Alessandria
ABSTRACT
Background: The lack of facile methods for the specific characterization of malignant thyroid nodules makes the diagnosis of thyroid cancer (TC) challenging. Due to its restricted expression in such nodules, the cell-associated lectin galectin-3 (Gal3) has emerged as a marker for TC with growing interest for in vivo imaging as well as targeted radionuclide therapy. To accelerate translation into clinical application, we have developed a cognate chimeric human antigen-binding fragment (Fab) derived from the rat anti-Gal3 monoclonal antibody M3/38. Methods: The variable immunoglobulin (Ig) light and heavy chain sequences were cloned from the hybridoma cell line, and the corresponding Fab carrying human IgG1/κ constant genes was functionally produced in the periplasm of Escherichia coli and purified to homogeneity. To moderately prolong its plasma half-life and, thus, increase tumor uptake, the recombinant Fab was fused with a long disordered amino acid chain comprising in total 200 Pro, Ala, and Ser residues (PASylation). This novel tracer was subjected to in vitro characterization and in vivo validation by using two thyroid cancer orthotopic murine models. To this end, the αGal3-Fab-PAS200 was conjugated with deferoxamine (Dfo), labeled with 89Zr under mild conditions and tested for binding on TC cell lines. Athymic nude mice were inoculated either with FRO82-1 or with CAL62 tumor cells into the left thyroid lobe. After intravenous injection with ∼3.0 MBq of 89Zr-Dfo-PAS200-Fab, these mice were subjected to positron emission tomography (PET)/computed tomography imaging followed by quantification of tumor accumulation and immunohistochemical analysis. Results: The αGal3-Fab-PAS200 revealed high affinity toward the recombinant Gal3 antigen, with a dissociation constant ≤1 nM as measured via enzyme-linked immunosorbent assay, surface plasmon resonance spectroscopy, and radioactive cell binding assay. The in vivo Gal3-targeting by the 89Zr(IV)-labeled protein tracer, as investigated by immuno-PET, demonstrated highly selective and fast accumulation in orthotopically implanted tumors, with strong contrast images achieved 24 hours postinjection, and no uptake in the tumor-free thyroid lobe, as also confirmed by biodistribution studies. Conclusions: The chimeric αGal3 89Zr-Dfo-PAS200-Fab tracer exhibits selective accumulation in the tumor-bearing thyroid lobe of xenograft mice. Thus, this novel radioactive probe offers potential to change TC management, in addition to current diagnostic procedures, and to reduce unnecessary thyroidectomies.