MilliporeSigma
  • Discovery of new tetracyclic tetrahydrofuran derivatives as potential broad-spectrum psychotropic agents.
Discovery of new tetracyclic tetrahydrofuran derivatives as potential broad-spectrum psychotropic agents.
Journal of medicinal chemistry (2005-03-18)
Javier Fernández, José M Alonso, José I Andrés, José M Cid, Adolfo Díaz, Laura Iturrino, Pilar Gil, Anton Megens, Victor K Sipido, Andrés A Trabanco
ABSTRACT
A series of novel tetracyclic tetrahydrofuran derivatives was prepared and evaluated for its potential psychotropic properties. In vitro affinities for multiple dopaminergic, serotonergic, alpha-adrenergic, and histamine receptors and for the norepinephrine transporter as well as the ED(50) values obtained in some in vivo assays for antipsychotic, anxiolytic, and antidepressant potential are reported. Compounds (-)-1, (-)-8d, and (+)-8d have been identified as potential broad-spectrum psychotropic agents.