MilliporeSigma
  • Heme deficiency sensitizes yeast cells to oxidative stress induced by hydroxyurea.
Heme deficiency sensitizes yeast cells to oxidative stress induced by hydroxyurea.
The Journal of biological chemistry (2017-04-06)
Amanpreet Singh, Yong-Jie Xu
ABSTRACT
Hydroxyurea (HU) has a long history of clinical and scientific use as an antiviral, antibacterial, and antitumor agent. It inhibits ribonucleotide reductase and reversibly arrests cells in S phase. However, high concentrations or prolonged treatment with low doses of HU can cause cell lethality. Although the cytotoxicity of HU may significantly contribute to its therapeutic effects, the underlying mechanisms remain poorly understood. We have previously shown that HU can induce cytokinesis arrest in the erg11-1 mutant of fission yeast, which has a partial defect in the biosynthesis of fungal membrane sterol ergosterol. Here, we report the identification of a new mutant in heme biosynthesis, hem13-1, that is hypersensitive to HU. We found that the HU hypersensitivity of the hem13-1 mutant is caused by oxidative stress and not by replication stress or a defect in cellular response to replication stress. The mutation is hypomorphic and causes heme deficiency, which likely sensitizes the cells to the HU-induced oxidative stress. Because the heme biosynthesis pathway is highly conserved in eukaryotes, this finding, as we show in our separate report, may help to expand the therapeutic spectrum of HU to additional pathological conditions.