MilliporeSigma
  • Rational design of non-nucleoside, potent, and orally bioavailable adenosine deaminase inhibitors: predicting enzyme conformational change and metabolism.
Rational design of non-nucleoside, potent, and orally bioavailable adenosine deaminase inhibitors: predicting enzyme conformational change and metabolism.
Journal of medicinal chemistry (2005-07-22)
Tadashi Terasaka, Kiyoshi Tsuji, Takeshi Kato, Isao Nakanishi, Takayoshi Kinoshita, Yasuko Kato, Masako Kuno, Takeshi Inoue, Kohichiro Tanaka, Katsuya Nakamura
ABSTRACT
From metabolic considerations and prediction of an inhibitor-induced conformational change, novel adenosine deaminase (ADA) inhibitors with improved activities and oral bioavailability have been developed on the basis of our originally designed non-nucleoside ADA inhibitors. They demonstrated in vivo efficacy in models of inflammation and lymphoma. Furthermore, X-ray crystal structure analysis has revealed a novel induced fit to ADA.
Sigma-Aldrich
Adenine 9-β-D-arabinofuranoside, ≥99%