MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • A chiral benzoquinolizine-2-carboxylic acid arginine salt active against vancomycin-resistant Staphylococcus aureus.
A chiral benzoquinolizine-2-carboxylic acid arginine salt active against vancomycin-resistant Staphylococcus aureus.
Journal of medicinal chemistry (2005-08-05)
Noel J de Souza, Shrikant V Gupte, Prasad K Deshpande, Vijaya N Desai, Satish B Bhawsar, Ravindra D Yeole, Milind C Shukla, Jacob Strahilevitz, David C Hooper, Bülent Bozdogan, Peter C Appelbaum, Michael R Jacobs, Nitin Shetty, Mahesh V Patel, Rasendrakumar Jha, Habil F Khorakiwala
ABSTRACT
There is an urgent medical need for novel antibacterial agents to treat hospital infections, specially those caused by multidrug-resistant Gram-positive pathogens. The need may also be fulfilled by either exploring antibacterial agents having new mechanism of action or expanding known classes of antibacterial drugs. The paper describes a new chemical entity, compound 21, derived from hitherto little known "floxacin". The choice of the entity was made from a series of synthesized prodrugs and salts of the active chiral benzoquinolizine carboxylic acid, S-(-)-nadifloxacin. The chemistry, physicochemical characteristics, and essential bioprofile of 21 qualifies it for serious consideration as a novel drug entity against hospital infections of multi-drug-resistant Staphylococcus aureus, and its progress up to clinical phase I trials in humans is described.
Product Description
Supelco
Norfloxacin, analytical standard, ≥98% (TLC)
Supelco
Norfloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)