MilliporeSigma
  • A discriminant model constructed by the support vector machine method for HERG potassium channel inhibitors.
A discriminant model constructed by the support vector machine method for HERG potassium channel inhibitors.
Bioorganic & medicinal chemistry letters (2005-05-25)
Motoi Tobita, Tetsuo Nishikawa, Renpei Nagashima
ABSTRACT
HERG attracts attention as a risk factor for arrhythmia, which might trigger torsade de pointes. A highly accurate classifier of chemical compounds for inhibition of the HERG potassium channel is constructed using support vector machine. For two test sets, our discriminant models achieved 90% and 95% accuracy, respectively. The classifier is even applied for the prediction of cardio vascular adverse effects to achieve about 70% accuracy. While modest inhibitors are partly characterized by properties linked to global structure of a molecule including hydrophobicity and diameter, strong inhibitors are exclusively characterized by properties linked to substructures of a molecule.
Product Description
Supelco
Ofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration), meets EP testing specifications
Supelco
Codeine monohydrate, analytical standard
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Terfenadine
Sigma-Aldrich
Nitrendipine, >95%, powder
Supelco
(−)-Nicotine solution, 1.0 mg/mL, analytical standard, for drug analysis
Supelco
(−)-Nicotine, PESTANAL®, analytical standard
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid