MilliporeSigma
  • Synthesis of benzoyl phenyl benzoates as effective inhibitors for phospholipase A2 and hyaluronidase enzymes.
Synthesis of benzoyl phenyl benzoates as effective inhibitors for phospholipase A2 and hyaluronidase enzymes.
Bioorganic & medicinal chemistry letters (2005-07-05)
Shaukath Ara Khanum, Satish Kumar Murari, Bannikuppe Sannanaik Vishwanth, Sheena Shashikanth
ABSTRACT
Benzoylation of (hydroxy phenyl) phenyl methanone 2a-g to benzoyl phenyl benzoates 4a-g, a benzophenone analogue, was achieved in good yield. All the newly synthesized compounds were evaluated for their phospholipase A2 [E.C. 3.1.1.4] and hyaluronidase [E.C. 3.2.1.35] enzyme inhibitory activity in snake venom as source and their structure-activity relationship with respect to different groups is reported for the first time. The in vitro PLA2 enzyme inhibitory activity and in vivo anti-inflammatory activity studies of benzoyl phenyl benzoates are illustrated.
Product Description
Supelco
Ursolic acid, analytical standard
Sigma-Aldrich
Ursolic acid, ≥90%