MilliporeSigma
  • Inclusion complexation of tolbutamide with ?-cyclodextrin and hydroxypropyl-?-cyclodextrin
Inclusion complexation of tolbutamide with ?-cyclodextrin and hydroxypropyl-?-cyclodextrin
Journal of psychosomatic obstetrics and gynaecology (1996)
Veiga F, et al.