MilliporeSigma
  • Metformin protects rotenone-induced dopaminergic neurodegeneration by reducing lipid peroxidation.
Metformin protects rotenone-induced dopaminergic neurodegeneration by reducing lipid peroxidation.
Pharmacological reports : PR (2020-03-25)
Gul Ozbey, Dilara Nemutlu-Samur, Hande Parlak, Sendegul Yildirim, Mutay Aslan, Gamze Tanriover, Aysel Agar
ABSTRACT
Metformin, a widely prescribed antidiabetic drug, has been suggested to have a neuroprotective effect on 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP)-induced neurotoxicity in mice. In this study, we investigated the neuroprotective potential of metformin against rotenone-induced dopaminergic neuron damage and its underlying mechanisms. C57BL/6 mice were given saline or rotenone (2.5 mg/kg/day, ip) injection for 10 days. Metformin treatment (300 mg/kg/day, ip) was started concurrently with rotenone administration and continued for 10 days. The neuroprotective effect of metformin on rotenone-induced dopaminergic toxicity was assessed by tyrosine hydroxylase (TH), cleaved caspase-3 and α-synuclein immunohistochemistry in substantia nigra (SN). SN tissues were extracted for biochemical analysis. Malondialdehyde (MDA) and 4-hydroxynonenal (4-HNE) protein levels were measured by spectrophotometric assay. We found that metformin treatment attenuated the rotenone-induced loss of TH+ neurons in the SN. Additionally, metformin significantly decreased the rotenone-induced increase of cleaved caspase-3 and α-synuclein accumulation in the SN; however, there was no difference in motor behaviours between the experimental groups. Meanwhile, the levels of MDA and 4-HNE in SN were significantly reduced in the rotenone-metformin group compared to the rotenone group. Results showed that metformin treatment attenuated dopaminergic neuron loss in SN induced by rotenone by decreasing lipid peroxidation.