MilliporeSigma
ClaI. a new restriction endonuclease from Caryophanon latum L.
Nucleic acids research (1981-10-10)
H Mayer, R Grosschedl, H Schütte, G Hobom
ABSTRACT
From Caryophanon latum L site specific restriction endonuclease (ClaI) has been purified, which recognises tha DNA hexanucleotide palindrome 5'-A-T-C-G-A-T-3'. Staggered cleavage generates DNA restriction fragments with 5'-terminal pCG extensions. A CLaI map of bacteriophage lambda has been determined, which indicates cleavage inhibition due to adenine methylation at over lapping ClaI-GATC recognition sequences. Plasmid pBR322 is cut only once, in the tetracycline promoter region, and can, therefore, be used as a vector system for cloning fragments derived from ClaI digestions, and in addition for fragments generated by TaqI, HpaII, and several other enzymes.