MilliporeSigma
  • Synthesis of benz[d]indeno[1,2-b]pyran-5,11-diones: versatile intermediates for the design and synthesis of topoisomerase I inhibitors.
Synthesis of benz[d]indeno[1,2-b]pyran-5,11-diones: versatile intermediates for the design and synthesis of topoisomerase I inhibitors.
Bioorganic & medicinal chemistry letters (2006-01-31)
Andrew Morrell, Smitha Antony, Glenda Kohlhagen, Yves Pommier, Mark Cushman
ABSTRACT
A method has been developed that relies on a two-step, one-pot condensation between phthalide and 2-carboxybenzaldehydes to provide benz[d]indeno[1,2-b]pyran-5,11-diones in a multi-gram fashion. Treatment of these compounds with a primary amine allows rapid access to various N-substituted indenoisoquinolines, whose in vitro anticancer activity and topoisomerase I inhibition have been evaluated.
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder