MilliporeSigma
  • The molecular basis for the selection of captopril cis and trans conformations by angiotensin I converting enzyme.
The molecular basis for the selection of captopril cis and trans conformations by angiotensin I converting enzyme.
Bioorganic & medicinal chemistry letters (2006-08-08)
Andreas G Tzakos, Nawazish Naqvi, Konstantinos Comporozos, Roberta Pierattelli, Vassiliki Theodorou, Ahsan Husain, Ioannis P Gerothanassis
ABSTRACT
Enzyme-inhibitor recognition is considered one of the most fundamental aspects in the area of drug discovery. However, the molecular mechanism of this recognition process (induced fit or prebinding and adaptive selection among multiple conformers) in several cases remains unexplored. In order to shed light toward this step of the recognition process in the case of human angiotensin I converting enzyme (hACE) and its inhibitor captopril, we have established a novel combinatorial approach exploiting solution NMR, flexible docking calculations, mutagenesis, and enzymatic studies. We provide evidence that an equimolar ratio of the cis and trans states of captopril exists in solution and that the enzyme selects only the trans state of the inhibitor that presents architectural and stereoelectronic complementarity with its substrate binding groove.
Sigma-Aldrich
Captopril, meets USP testing specifications
Sigma-Aldrich
Captopril, ≥98% (HPLC), powder