MilliporeSigma
  • ScrFI: a new sequence-specific endonuclease from Streptococcus cremoris.
ScrFI: a new sequence-specific endonuclease from Streptococcus cremoris.
Nucleic acids research (1982-12-20)
G F Fitzgerlad, C Daly, L R Brown, T R Gingeras
ABSTRACT
A novel sequence-specific endonuclease has been isolated from Streptococcus cremoris F. ScrFI recognises the sequence: (formula; see text) and cleaves as indicated by the arrow ( ). It is the first enzyme to recognise this sequence and the first endonuclease reported from the lactic streptococci used in dairy fermentations.