MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Synthesis, characterization, antibacterial activity, and interaction with DNA of the vanadyl-enrofloxacin complex.
Synthesis, characterization, antibacterial activity, and interaction with DNA of the vanadyl-enrofloxacin complex.
Bioorganic & medicinal chemistry letters (2007-01-02)
Eleni K Efthimiadou, Nikos Katsaros, Alexandra Karaliota, George Psomas
ABSTRACT
The neutral mononuclear vanadyl complex with the quinolone antibacterial drug enrofloxacin has been prepared and characterized with physicochemical and spectroscopic techniques and molecular mechanics calculations. The interaction of the complex with calf-thymus DNA has also been investigated and the antimicrobial activity has been evaluated against three different microorganisms.
Product Description
Supelco
Enrofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Enrofloxacin, ≥99.0%