MilliporeSigma
  • Mutant myogenin promoter-controlled oncolytic adenovirus selectively kills PAX3-FOXO1-positive rhabdomyosarcoma cells.
Mutant myogenin promoter-controlled oncolytic adenovirus selectively kills PAX3-FOXO1-positive rhabdomyosarcoma cells.
Translational oncology (2020-12-19)
Hideki Yoshida, Mizuho Sato-Dahlman, Praveensingh Hajeri, Kari Jacobsen, Lisa Koodie, Chikako Yanagiba, Ryan Shanley, Masato Yamamoto
ABSTRACT
The PAX3-FOXO1 fusion gene functions as a transactivator and increases expression of many cancer-related genes. These lead to metastases and other unfavorable outcomes for alveolar rhabdomyosarcoma (ARMS) patients. In order to target ARMS with the PAX3-FOXO1 transactivator, we developed an Oncolytic Adenovirus (OAd) regulated by the myogenin (pMYOG) promoter with a mutation in the Myocyte Enhancer Factor-2 binding site (mMEF2) in this study. The expression of MYOG in the two RMS cell lines (Rh30; PAX3-FOXO1-positive, RD; PAX3-FOXO1-negative) is about 1,000 times higher than normal skeletal muscle cell (SkMC). Ad5/3-pMYOG(S)-mMEF2 (short-length pMYOG-controlled OAd with mMEF2) showed strong replication and cytocidal effect in Rh30, but to a much lesser extent in RD. Ad5/3-pMYOG(S) (pMYOG-controlled OAd with native pMYOG) showed similar effects in RD and Rh30. Neither virus killed SkMC, indicating that Ad5/3-pMYOG(S)-mMEF2 selectively replicates and kills cells with PAX3-FOXO1. Additionally, Ad5/3-pMYOG(S)-mMEF2 showed replication and spread in vitro as well as tumor growth suppression and intratumoral viral spread in vivo, selectively in Rh30 not in RD. Our findings revealed that Ad5/3-pMYOG(S)-mMEF2 shows a promise as a safe and potent therapy to improve treatment in PAX3-FOXO1-positive ARMSs.
Sigma-Aldrich
Anti-Adenovirus Antibody, FITC-conjugated, Chemicon®, from goat