MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Molecular modeling of some 1H-benzimidazole derivatives with biological activity against Entamoeba histolytica: a comparative molecular field analysis study.
Molecular modeling of some 1H-benzimidazole derivatives with biological activity against Entamoeba histolytica: a comparative molecular field analysis study.
Bioorganic & medicinal chemistry (2006-11-01)
Fabian López-Vallejo, José Luis Medina-Franco, Alicia Hernández-Campos, Sergio Rodríguez-Morales, Lilián Yépez, Roberto Cedillo, Rafael Castillo
ABSTRACT
Comparative molecular field analysis (CoMFA) was performed on a set of 1H-benzimidazole derivatives. Molecular modeling and 3D-QSAR were employed to determine the tautomeric form that would probably fit a target receptor in Entamoeba histolytica. CoMFA results suggest that the antiamoebic activity is favored with steric bulk at position 5 of the benzimidazole ring and low electron density on the group at position 2. To the best of our knowledge this is the first 3D-QSAR study performed for benzimidazoles as antiamoebic agents. The CoMFA models derived will be very valuable to design new and more potent compounds against E. histolytica.
Sigma-Aldrich
2-Aminobenzimidazole, 97%
Sigma-Aldrich
2-Methylbenzimidazole, 98%
Sigma-Aldrich
Benzimidazole, 98%