MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • 'Bridged' stilbene derivatives as selective cyclooxygenase-1 inhibitors.
'Bridged' stilbene derivatives as selective cyclooxygenase-1 inhibitors.
Bioorganic & medicinal chemistry (2007-07-03)
Norbert Handler, Gerda Brunhofer, Christian Studenik, Klaus Leisser, Walter Jaeger, Stephanie Parth, Thomas Erker
ABSTRACT
Resveratrol ((E)-3,4',5-trihydroxy-stilbene), a phytoalexin found in various plants, shows non-selective cyclooxygenase-1 (COX-1) and cyclooxygenase-2 (COX-2) inhibition. In order to find more selective COX inhibitors a series of bridged stilbene derivatives was synthesized and evaluated for their ability to inhibit both COX-1 and COX-2 in vitro. The compounds showed a high rate of COX-1 inhibition with the most potent compounds exhibiting submicromolar IC(50) values and high selectivity indices. A prediction model for COX-inhibiting activity was also developed using the classical LIE approach resulting in consistent docking data for our molecule sample. Phenyl substituted 1,2-dihydronaphthalene derivatives and 1H-indene derivatives therefore represent a novel class of highly selective COX-1 inhibitors and land promising candidates for in vivo studies.
Product Description
Supelco
Resveratrol, analytical standard
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
1,2-Dihydronaphthalene, 95%
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%