MilliporeSigma
  • Norcantharimides, synthesis and anticancer activity: Synthesis of new norcantharidin analogues and their anticancer evaluation.
Norcantharimides, synthesis and anticancer activity: Synthesis of new norcantharidin analogues and their anticancer evaluation.
Bioorganic & medicinal chemistry (2007-07-04)
Timothy A Hill, Scott G Stewart, Stephen P Ackland, Jayne Gilbert, Benjamin Sauer, Jennette A Sakoff, Adam McCluskey
ABSTRACT
A range of amines was reacted with norcantharidin (2) to provide the corresponding norcantharimides (9-43). Treatment of norcantharidin with allylamine afforded the corresponding allyl-norcantharimide (20) which was amenable to epoxidation (mCPBA, 22) and subsequent ring opening (MeOH/H(+); 23) or alternatively, osmylation (OsO(4)/NMO; 24). These simple synthetic modifications of 2 facilitated the development of a novel series of norcantharimides displaying modest to good broad spectrum cytotoxicity against HT29 and SW480 (colorectal carcinoma); MCF-7 (breast adenocarcinoma); A2780 (ovarian carcinoma); H460 (lung carcinoma); A431 (epidermoid carcinoma); DU145 (prostate carcinoma); BE2-C (neuroblastoma); and SJ-G2 (glioblastoma). Analogues possessing a C(10), C(12) or C(14) alkyl chain or a C(12) linked bis-norcantharimide displayed the highest levels of cytotoxicity.
Sigma-Aldrich
Cantharidin
Pricing and availability is not currently available.