MilliporeSigma
  • RET fusion genes are associated with chronic myelomonocytic leukemia and enhance monocytic differentiation.
RET fusion genes are associated with chronic myelomonocytic leukemia and enhance monocytic differentiation.
Leukemia (2012-04-20)
P Ballerini, S Struski, C Cresson, N Prade, S Toujani, C Deswarte, S Dobbelstein, A Petit, H Lapillonne, E-F Gautier, C Demur, E Lippert, P Pages, V Mansat-De Mas, J Donadieu, F Huguet, N Dastugue, C Broccardo, C Perot, E Delabesse
ABSTRACT
Myeloproliferative neoplasms are frequently associated with aberrant constitutive tyrosine kinase (TK) activity resulting from chimaeric fusion genes or point mutations such as BCR-ABL1 or JAK2 V617F. We report here the cloning and functional characterization of two novel fusion genes BCR-RET and FGFR1OP-RET in chronic myelomonocytic leukemia (CMML) cases generated by two balanced translocations t(10;22)(q11;q11) and t(6;10)(q27;q11), respectively. The two RET fusion genes leading to the aberrant activation of RET, are able to transform hematopoietic cells and skew the hematopoietic differentiation program towards the monocytic/macrophage lineage. The RET fusion genes seem to constitutively mimic the same signaling pathway as RAS mutations frequently involved in CMML. One patient was treated with Sorafenib, a specific inhibitor of the RET TK function, and demonstrated cytological and clinical remissions.
Sigma-Aldrich
Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody produced in mouse, clone MAPK-YT, ascites fluid
Sigma-Aldrich
Anti-c-Abl (Ab-3) Mouse mAb (24-21), liquid, clone 24-21, Calbiochem®