MilliporeSigma
  • Specialized metabolites of the United States lichen Niebla homalea and their antiproliferative activities.
Specialized metabolites of the United States lichen Niebla homalea and their antiproliferative activities.
Phytochemistry (2020-10-26)
Yan Zhang, Choon Yong Tan, Richard W Spjut, James R Fuchs, A Douglas Kinghorn, Liva Harinantenaina Rakatondraibe, Yan Zhang, Choon Yong Tan, Richard W Spjut, James R Fuchs, A Douglas Kinghorn, Liva Harinantenaina Rakatondraibe
ABSTRACT
Three undescribed stictanes, nieblastictanes A-C, two flavicanes, nieblaflavicanes A and B, together with three already reported stictanes, along with the known compounds (+)-usnic acid, sekikaic acid, divaricatic acid, and divaricatinic acid methyl ester were isolated from an ethyl acetate extract of the western North American lichen Niebla homalea. The structures of the new and known compounds were established by spectroscopic methods including nuclear magnetic resonance spectroscopy, mass spectrometry and electronic circular dichroism. Among the compounds isolated, usnic acid exhibited moderately potent antiproliferative activities against the A2780 ovarian (IC50 3.8 μM) and MCF-7 breast cancer (IC50 6.8 μM) cell lines. A plausible mode of formation of the chlorine-containing compound nieblastictane C is provided and the contribution of the isolated compounds to the chemotaxonomy of United States lichen species of the genus Niebla is also discussed.
Sigma-Aldrich
Round crystal window, KBr, diam. × thickness 25 mm × 5 mm