MilliporeSigma
  • A mutual prodrug ester of GABA and perphenazine exhibits antischizophrenic efficacy with diminished extrapyramidal effects.
A mutual prodrug ester of GABA and perphenazine exhibits antischizophrenic efficacy with diminished extrapyramidal effects.
Journal of medicinal chemistry (2008-03-28)
Abraham Nudelman, Irit Gil-Ad, Nava Shpaisman, Igor Terasenko, Hanna Ron, Kinneret Savitsky, Yona Geffen, Abraham Weizman, Ada Rephaeli
ABSTRACT
The perphenazine and fluphenazine GABA esters 3 and 4 evaluated in rat models for antipsychotic activity displayed a significant decrease of catalepsy associated with increased prolactin blood levels. Efficacy was evaluated in the d-amphetamine-induced hyperactivity model, where perphenazine abolished hyperactivity and induced sedation and catalepsy, whereas 3 reduced hyperactivity without sedation or catalepsy. Thus, 3 (BL-1020) constitutes a prototype of novel antipsychotics possessing GABAergic activity. A phase II study is in progress.
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%