MilliporeSigma
  • Evaluation of an epitope-blocking enzyme-linked immunosorbent assay for the diagnosis of West Nile virus infections in humans.
Evaluation of an epitope-blocking enzyme-linked immunosorbent assay for the diagnosis of West Nile virus infections in humans.
Clinical and vaccine immunology : CVI (2009-03-27)
M A Loroño-Pino, J A Farfan-Ale, B J Blitvich, J L Beebe, R G Jarman, B J Beaty
ABSTRACT
An epitope-blocking enzyme-linked immunosorbent assay (b-ELISA) was evaluated for the diagnosis of West Nile virus (WNV) infections in humans. Sera from patients diagnosed with WNV infections from an outbreak in 2003 in Colorado and from patients diagnosed with dengue virus infections from Mexico and Thailand were tested with the b-ELISA. The b-ELISAs were performed using the WNV-specific monoclonal antibody (MAb) 3.1112G and the flavivirus-specific MAb 6B6C-1. Although the WNV-specific b-ELISA was effective in diagnosing WNV infections in humans from Colorado, it was not efficacious for diagnosing WNV infections in serum specimens from Mexico and Thailand. In serum specimens from patients from Colorado, the WNV b-ELISA and the WNV plaque reduction neutralization test showed an overall agreement of 91%. The sensitivity and specificity of the WNV b-ELISA were 89% and 92%, respectively, with a false-positive rate of 5%, based on receiver operating characteristic analysis. In contrast, false-positive rate results in specimens from the countries of Mexico and Thailand, where flaviviruses are endemic, were 79% and 80%, presumably due to the presence of antibodies resulting from previous dengue virus infections in Mexico and/or Japanese encephalitis virus infections or vaccination in Thailand. Thus, in regions where people have experienced previous or multiple flavivirus infections, the use of the b-ELISA for WNV diagnosis is contraindicated.