MilliporeSigma
ErbB2 induces Notch1 activity and function in breast cancer cells.
Clinical and translational science (2008-09-01)
Jaime Lindsay, Xuanmao Jiao, Toshiyuki Sakamaki, Mathew C Casimiro, Lawrence A Shirley, Thai H Tran, Xiaoming Ju, Manran Liu, Zhiping Li, Chenguang Wang, Sanjay Katiyar, Mahadev Rao, Kathleen G Allen, Robert I Glazer, Changhui Ge, Pamela Stanley, Michael P Lisanti, Hallgeir Rui, Richard G Pestell
ABSTRACT
The ErbB2 (Her2/neu epidermal growth receptor family) oncogene is overexpressed in 30% to 40% of human breast cancers. Cyclin D1 is the regulatory subunit of the holoenzyme that phosphorylates and inactivates the retinoblastoma (pRb) tumor suppressor and is an essential downstream target of ErbB2-induced tumor growth. Herein, we demonstrate that ErbB2 induces the activity of the Notch signaling pathway. ErbB2 induction of DNA synthesis, contact-independent growth, and mammosphere induction required Notch1. ErbB2-induced cyclin D1 and cyclin D1 expression was suficient to induce Notch1 activity, and conversely, genetic deletion of Notch1 in mammary epithelial cells using foxed Notch (Notch(fl/fl)) mice demonstrated that cyclin D1 is induced by Notch1. Genetic deletion of cyclin D1 or small interfering RNA (siRNA) to cyclin D1-reduced Notch1 activity and reintroduction of cyclin D1 into cyclin D1-deficient cells restored Notch1 activity through the inhibition of Numb, an endogenous inhibitor of Notch1 activity. Thus, cyclin D1 functions downstream as a genetic target of Notch1, amplifies Notch1 activity by repressing Numb, and identifies a novel pathway by which ErbB2 induces Notch1 activity via the induction of cyclin D1.
Sigma-Aldrich
γ-Secretase Inhibitor X, InSolution, ≥90%, 1 mM