MilliporeSigma
  • Synthesis and biological evaluation of N-mercaptoacylproline and N-mercaptoacylthiazolidine-4-carboxylic acid derivatives as leukotriene A4 hydrolase inhibitors.
Synthesis and biological evaluation of N-mercaptoacylproline and N-mercaptoacylthiazolidine-4-carboxylic acid derivatives as leukotriene A4 hydrolase inhibitors.
Bioorganic & medicinal chemistry letters (2008-08-05)
Hiroshi Enomoto, Yuko Morikawa, Yurika Miyake, Fumio Tsuji, Maki Mizuchi, Hiroshi Suhara, Ken-ichi Fujimura, Masato Horiuchi, Masakazu Ban
ABSTRACT
We studied the synthetic modification of structurally similar N-mercaptoacyl-L-proline and (4R)-N-mercaptoacylthiazolidine-4-carboxylic acid to obtain potent leukotriene A(4) (LTA(4)) hydrolase inhibitors. An N-mercaptoacyl group, (2S)-3-mercapto-2-methylpropionyl group, was effective for both scaffolds. Additional introduction of a large substituent such as 4-isopropylbenzylthio (3f), 4-tert-butylbenzylthio (3l) or 4-cyclohexylbenzylthio group (3m) with (S)-configuration at the C(4) position of proline yielded much more potent LTA(4) hydrolase inhibitors (IC(50); 52, 31, and 34 nM, respectively) than captopril (IC(50); 630,000 nM).
Sigma-Aldrich
Captopril, meets USP testing specifications
Sigma-Aldrich
Captopril, ≥98% (HPLC), powder