MilliporeSigma
  • Glutathione peroxidase protects against peroxynitrite-mediated oxidations. A new function for selenoproteins as peroxynitrite reductase.
Glutathione peroxidase protects against peroxynitrite-mediated oxidations. A new function for selenoproteins as peroxynitrite reductase.
The Journal of biological chemistry (1997-11-05)
H Sies, V S Sharov, L O Klotz, K Briviba
ABSTRACT
There is a requirement for cellular defense against excessive peroxynitrite generation to protect against DNA strand breaks and mutations and against interference with protein tyrosine-based signaling and other protein functions due to formation of 3-nitrotyrosine. Here, we demonstrate a role of selenium-containing enzymes catalyzing peroxynitrite reduction using glutathione peroxidase (GPx) as an example. GPx protected against the oxidation of dihydrorhodamine 123 by peroxynitrite more effectively than ebselen (2-phenyl-1,2-benzisoselenazol-3(2H)-one), a selenoorganic compound exhibiting a high second-order rate constant for the reaction with peroxynitrite, 2 x 10(6) M-1 s-1. Carboxymethylation of selenocysteine in GPx by iodoacetate led to the loss of "classical" glutathione peroxidase activity but maintained protection against peroxynitrite-mediated oxidation. The maintenance of protection by GPx against peroxynitrite requires GSH as reductant. When peroxynitrite was infused to maintain a 0.2 microM steady-state concentration, GPx in the presence of GSH, but neither GPx nor GSH alone, effectively inhibited the hydroxylation of benzoate by peroxynitrite. Under these steady-state conditions peroxynitrite did not cause the loss of classical GPx activity. GPx, like selenomethionine, protected against protein 3-nitrotyrosine formation in human fibroblast lysates, shown in Western blots. The formation of nitrite rather than nitrate from peroxynitrite was enhanced by GPx or by selenomethionine. The results demonstrate a novel function of GPx and potentially of other selenoproteins containing selenocysteine or selenomethionine, in the GSH-dependent maintenance of a defense line against peroxynitrite-mediated oxidations, as a peroxynitrite reductase.
Sigma-Aldrich
Seleno-L-methionine, ≥98% (TLC), powder