MilliporeSigma
Bilirubin in clinical practice: a review.
Liver international : official journal of the International Association for the Study of the Liver (2008-04-25)
Johan Fevery
ABSTRACT
Bilirubin is an endogenous compound that can be toxic under certain conditions but, on the other hand, mild unconjugated hyperbilirubinaemia might protect against cardiovascular diseases and tumour development. Serum bilirubin levels are often enhanced under a variety of clinical conditions. These are discussed and the mechanisms are outlined.
Product Description
Millipore
Bilirubin, Principal pigment of bile and one of the major end products of hemoglobin decomposition.