MilliporeSigma
  • 2-Substituted 4-, 5-, and 6-[(1E)-3-oxo-3-phenylprop-1-en-1-yl]pyridazin-3(2H)-ones and 2-substituted 4,5-bis[(1E)-3-oxo-3-phenylprop-1-en-1-yl]pyridazin-3(2H)-ones as potent platelet aggregation inhibitors: design, synthesis, and SAR studies.
2-Substituted 4-, 5-, and 6-[(1E)-3-oxo-3-phenylprop-1-en-1-yl]pyridazin-3(2H)-ones and 2-substituted 4,5-bis[(1E)-3-oxo-3-phenylprop-1-en-1-yl]pyridazin-3(2H)-ones as potent platelet aggregation inhibitors: design, synthesis, and SAR studies.
Bioorganic & medicinal chemistry letters (2007-12-07)
Caroline Meyers, Matilde Yáñez, Abdelaziz Elmaatougi, Tom Verhelst, Alberto Coelho, Nuria Fraiz, Guy L F Lemière, Xerardo García-Mera, Reyes Laguna, Ernesto Cano, Bert U W Maes, Eddy Sotelo
ABSTRACT
A set of regioisomeric 2-substituted pyridazin-3(2H)-ones containing a 3-oxo-3-phenylprop-1-en-1-yl fragment at either position 4, 5 or 6 and 2-substituted pyridazin-3(2H)-ones containing the same fragment both at positions 4 and 5 have been synthesized and evaluated as antiplatelet agents. The study allows the identification of a new highly potent platelet aggregation inhibitor (4c).
Sigma-Aldrich
trans-Chalcone, 97%