MilliporeSigma
  • Tetrahydrobiopterin synthesis. An absolute requirement for cytokine-induced nitric oxide generation by vascular smooth muscle.
Tetrahydrobiopterin synthesis. An absolute requirement for cytokine-induced nitric oxide generation by vascular smooth muscle.
The Journal of biological chemistry (1992-12-25)
S S Gross, R Levi
ABSTRACT
Nitric oxide (NO) synthesis is induced in vascular smooth muscle cells by lipopolysaccharide (LPS) where it appears to mediate a variety of vascular dysfunctions. In some cell types tetrahydrobiopterin (BH4) synthesis has also been found to be induced by cytokines. Because BH4 is a cofactor for NO synthase, we investigated whether BH4 synthesis is required for LPS-induced NO production in rat aortic smooth muscle cells (RASMC). The total biopterin content (BH4 and more oxidized states) of untreated RASMC was below our limit of detection. However, treatment with LPS caused a significant rise in biopterin levels and an induction of NO synthesis; both effects of LPS were markedly potentiated by interferon-gamma. 2,4-Diamino-6-hydroxypyrimidine (DAHP), a selective inhibitor of GTP cyclohydrolase I, the rate-limiting enzyme for de novo BH4 synthesis, completely abolished the elevated biopterin levels induced by LPS. DAHP also caused a concentration-dependent inhibition of LPS-induced NO synthesis. Inhibition of NO synthesis by DAHP was reversed by sepiapterin, an agent which circumvents the inhibition of biopterin synthesis by DAHP by serving as a substrate for BH4 synthesis via the pterin salvage pathway. The reversal by sepiapterin was overcome by methotrexate, an inhibitor of the pterin salvage pathway. Sepiapterin, and to a lesser extent BH4, dose-dependently enhanced LPS-induced NO synthesis, indicating that BH4 concentration limits the rate of NO production by LPS-activated RASMC. Sepiapterin also caused LPS-induced NO synthesis to appear with an abbreviated lag period phase, suggesting that BH4 availability also limits the onset of NO synthesis. In contrast to the stimulation of LPS-induced NO synthesis, observed when sepiapterin was given alone, sepiapterin became a potent inhibitor of NO synthesis in the presence of methotrexate. This is attributable to a direct inhibitory action of sepiapterin on GTP cyclohydrolase I, an activity which is only revealed after blocking the metabolism of sepiapterin to BH4. Further studies with sepiapterin, methotrexate, and N-acetylserotonin (an inhibitor of the BH4 synthetic enzyme, sepiapterin reductase) indicated that the BH4 is synthesized in RASMC predominantly from GTP; however, a lesser amount may derive from pterin salvage. We demonstrate that BH4 synthesis is an absolute requirement for induction of NO synthesis by LPS in vascular smooth muscle. Our findings also suggest that pterin synthesis inhibitors may be useful for the therapy of endotoxin- and cytokine-induced shock.