MilliporeSigma
  • Orally administered FPRL1 receptor agonist peptide MMK-1 inhibits etoposide-induced alopecia by a mechanism different from intraperitoneally administered MMK-1.
Orally administered FPRL1 receptor agonist peptide MMK-1 inhibits etoposide-induced alopecia by a mechanism different from intraperitoneally administered MMK-1.
Peptides (2005-09-27)
Takahiro Tsuruki, Masaaki Yoshikawa
ABSTRACT
Oral administration for 6 days of 100 mg/kg MMK-1, an agonist peptide selective for the FPRL1 receptor, suppressed alopecia induced by the anticancer drug etoposide in neonatal rats. The anti-alopecia effect of orally administered MMK-1 was not inhibited by pyrilamine or cimetidine, antagonists for histamine H1 and H2 receptors, respectively, which blocked the anti-alopecia effect of intraperitoneally administered MMK-1 at a dose of 10 mg/kg for 4 days. However, the anti-alopecia effect of orally administered MMK-1 was inhibited by indomethacin, an inhibitor of cyclooxygenase (COX), or AH-23848B, an antagonist of the EP4 receptor for prostaglandin (PG) E2, suggesting involvement of PGE2 release and the EP4 receptor in the oral MMK-1 anti-alopecia mechanism. The anti-alopecia effect of orally administered MMK-1 was also blocked by an inhibitor of nuclear factor-kappaB (NF-kappaB), pyrrolidine dithiocarbamate, suggesting that the oral anti-alopecia effect of MMK-1 may be mediated by activation of NF-kappaB. These results suggest that MMK-1 bound to FPRL1 receptor might suppress etoposide-induced apoptosis of hair follicle cells and alopecia by way of PGE2 release and NF-kappaB activation.
Sigma-Aldrich
MMK-1 trifluoroacetate, ≥95% (HPLC)