MilliporeSigma
  • Thiopyrano[2,3-e]indol-2-ones: angelicin heteroanalogues with potent photoantiproliferative activity.
Thiopyrano[2,3-e]indol-2-ones: angelicin heteroanalogues with potent photoantiproliferative activity.
Bioorganic & medicinal chemistry (2008-10-28)
Paola Barraja, Patrizia Diana, Alessandra Montalbano, Anna Carbone, Girolamo Cirrincione, Giampietro Viola, Alessia Salvador, Daniela Vedaldi, Francesco Dall'acqua
ABSTRACT
A new class of compounds, the thiopyrano[2,3-e]indol-2-ones, bioisosters of the angular furocoumarin angelicin, was synthesized with the aim of obtaining new photochemotherapeutic agents. In particular 7,8-dimethyl-thiopyranoindolone 6c s showed a remarkable phototoxicity and a great dose UVA dependence reaching IC(50) values at submicromolar level. This latter photoinduced a massive apoptosis and a remarkable photodamage to lipids and proteins. Although it did not intercalate DNA, it was able to cause photooxidation of DNA bases.
Product Description
Supelco
8-Methoxypsoralen, analytical standard