MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Activity of Tigecycline against Coryneform Bacteria of Clinical Interest and Listeria monocytogenes.
Activity of Tigecycline against Coryneform Bacteria of Clinical Interest and Listeria monocytogenes.
Antimicrobial agents and chemotherapy (2008-01-30)
Carlos Salas, Jorge Calvo, Luis Martínez-Martínez
ABSTRACT
The activities of tigecycline and eight other agents were evaluated against 220 coryneform bacteria and 42 Listeria monocytogenes isolates. All strains were inhibited by tigecycline at 0.5 mug/ml, except for 11 Corynebacterium striatum strains that were inhibited at 1 mug/ml. Tigecycline shows good in vitro activity against coryneform bacteria and L. monocytogenes.
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)