MilliporeSigma
  • Histone H2B monoubiquitination is a critical epigenetic switch for the regulation of autophagy.
Histone H2B monoubiquitination is a critical epigenetic switch for the regulation of autophagy.
Nucleic acids research (2017-02-10)
Su Chen, Yuanya Jing, Xuan Kang, Lu Yang, Da-Liang Wang, Wei Zhang, Lei Zhang, Ping Chen, Jian-Feng Chang, Xiao-Mei Yang, Fang-Lin Sun
ABSTRACT
Autophagy is an evolutionarily conserved cellular process that primarily participates in lysosome-mediated protein degradation. Although autophagy is a cytoplasmic event, how epigenetic pathways are involved in the regulation of autophagy remains incompletely understood. Here, we found that H2B monoubiquitination (H2Bub1) is down-regulated in cells under starvation conditions and that the decrease in H2Bub1 results in the activation of autophagy. We also identified that the deubiquitinase USP44 is responsible for the starvation-induced decrease in H2Bub1. Furthermore, the changes in H2Bub1 affect the transcription of genes involved in the regulation of autophagy. Therefore, this study reveals a novel epigenetic pathway for the regulation of autophagy through H2Bub1.
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
CpGenome Direct Prep Bisulfite Modification Kit (50 Reactions), The CpGenome Direct Prep Bisulfite Modification Kit allows bisulfite conversion directly from a variety of starting materials, including cultured cells, blood, fresh tissue & fixed tissue samples.