MilliporeSigma
  • A Renewable Source of Human Beige Adipocytes for Development of Therapies to Treat Metabolic Syndrome.
A Renewable Source of Human Beige Adipocytes for Development of Therapies to Treat Metabolic Syndrome.
Cell reports (2018-12-13)
Su Su, Anyonya R Guntur, Daniel C Nguyen, Shameem S Fakory, Chad C Doucette, Cassandra Leech, Humphrey Lotana, Matthew Kelley, Jaspreet Kohli, Julieta Martino, Sunder Sims-Lucas, Lucy Liaw, Calvin Vary, Clifford J Rosen, Aaron C Brown
ABSTRACT
Molecular- and cellular-based therapies have the potential to reduce obesity-associated disease. In response to cold, beige adipocytes form in subcutaneous white adipose tissue and convert energy stored in metabolic substrates to heat, making them an attractive therapeutic target. We developed a robust method to generate a renewable source of human beige adipocytes from induced pluripotent stem cells (iPSCs). Developmentally, these cells are derived from FOXF1+ mesoderm and progress through an expandable mural-like mesenchymal stem cell (MSC) to form mature beige adipocytes that display a thermogenically active profile. This includes expression of uncoupling protein 1 (UCP1) concomitant with increased uncoupled respiration. With this method, dysfunctional adipogenic precursors can be reprogrammed and differentiated into beige adipocytes with increased thermogenic function and anti-diabetic secretion potential. This resource can be used to (1) elucidate mechanisms that underlie the control of beige adipogenesis and (2) generate material for cellular-based therapies that target metabolic syndrome in humans.
Sigma-Aldrich
TGF-β RI Kinase Inhibitor VI, SB431542, TGF-β RI Kinase Inhibitor VI, SB431542, CAS 301836-41-9, is a cell-permeable inhibitor of SMAD2 phosphorylation. Inhibits the activity of ALK4 and ALK5 (IC50 = 140 nM and 94 nM, respectively).