MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Synthesis and biological evaluation of arylhydrazinocyanoacrylates and N-aryl pyrazolecarboxylates.
Synthesis and biological evaluation of arylhydrazinocyanoacrylates and N-aryl pyrazolecarboxylates.
Bioorganic & medicinal chemistry letters (2009-05-02)
Yuxiu Liu, Shaohua Liu, Yonghong Li, Haibin Song, Qingmin Wang
ABSTRACT
A series of arylhydrazino-substituted cyanoacrylates 3 and N-aryl pyrazolecarboxylates 6 were synthesized and their bioactivities were evaluated. Though compounds 3 were designed as herbicide, some of them showed fungicidal activity and anti-tumor activity. Some of the compounds 6 exhibited plant growth regulatory activity.
Sigma-Aldrich
3-Indoleacetic acid, suitable for plant cell culture, crystalline
Sigma-Aldrich
3-Indoleacetic acid, 98%
Supelco
3-Indoleacetic acid, PESTANAL®, analytical standard