MilliporeSigma
  • Use of the labscale tangential flow diafiltrator to remove tetrameric hemoglobin from polyhemoglobin, purify hemolysate, and concentrate polyhemoglobin.
Use of the labscale tangential flow diafiltrator to remove tetrameric hemoglobin from polyhemoglobin, purify hemolysate, and concentrate polyhemoglobin.
Artificial cells, blood substitutes, and immobilization biotechnology (2009-02-21)
Ada Stefanescu, Thomas Ming Swi Chang
ABSTRACT
The Labscale tangential flow diafiltration system in conjunction with polyethersulfone Biomax filters (both purchased from Millipore Inc.) were used in the following studies: (1) A crucial step in the preparation of polyhemoglobin (polyHb) is the removal of tetrameric hemoglobin (Hb), which can cause adverse side-effects. The efficiency for this depends on the integrity of the 100kDa filters. Those with lower integrity were not effective whereas those with a higher integrity of 0.075ml/min were more effective. A filter with an integrity of 0.075ml/min can reduce the initial 11.2% tetrameric Hb concentration to 1.3% using a flow rate of 0.2ml/min. Higher flow rate of 2.2ml/min was not as effective. (2) Filtration with a 100kDa filter was quick and efficient in separating stroma-free hemoglobin solutions from cellular debris from hemolysate. (3) Both 5kDa and 100kDa filters are efficient in concentrating hemoglobin solutions.
Sigma-Aldrich
Catalase Assay Kit, sufficient for ≥100 tests enzymatic, determination of catalase activity in tissues and cells