MilliporeSigma
Pas Kinase Deficiency Triggers Antioxidant Mechanisms in the Liver.
Scientific reports (2018-09-16)
P Dongil, A Pérez-García, V Hurtado-Carneiro, C Herrero-de-Dios, E Blazquez, E Alvarez, C Sanz
ABSTRACT
Metabolic dysfunction in the liver is the cause of numerous pathologies, which are associated with an altered redox state. PASK (PAS Domain Kinase) is a nutrient and bioenergetic sensor. We contend that PASK could act as an oxidative stress sensor in liver and/or control the metabolic balance, playing a role in the mitochondrial homeostasis. Using PASK-deficient mice, we observed that PASK deficiency promotes antioxidant response mechanisms: a lower production of ROS/RNS under non-fasting conditions, overexpression of genes coding to ROS-detoxifying enzymes and mitochondrial fusion proteins (MnSod Gpx, Mfn1 and Opa1), coactivator Ppargc1a, transcription factors (Pparg and FoxO3a) and deacetylase Sirt1. Also, under fasting conditions, PASK deficiency induced the overexpression of Ppargc1a, Ppara, Pparg, FoxO3a and Nrf2 leading to the overexpression of genes coding to antioxidant enzymes such as MnSOD, Cu/ZnSOD, GPx, HO1 and GCLm. Additionally, inducing PINK1 involved in cell survival and mitophagy. These changes kept ROS steady levels and improved the regenerative state. We suggest a new role for PASK as a controller of oxidative stress and mitochondrial dynamics in the liver. In fact, antioxidant response is PASK dependent. PASK-targeting could therefore be a good way of reducing the oxidative stress in order to prevent or treat liver diseases.
Sigma-Aldrich
Citrate Synthase Activity Assay Kit, Sufficient for 100 Colorimetric tests