MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Single-step PCR-based genetic sex determination of rat tissues and cells.
Single-step PCR-based genetic sex determination of rat tissues and cells.
BioTechniques (2017-05-23)
Pramod Dhakal, Michael J Soares
ABSTRACT
The advent of genome editing strategies has expanded the range of animal models available for gene manipulation and renewed research interest in the rat. Gender is a key variable for in vivo gene function analyses. Here, we present a simple PCR-based method to determine genetic sex in the rat.
Sigma-Aldrich
Extract-N-Amp Tissue PCR Kit, sufficient for 100 extractions, sufficient for 100 amplifications