MilliporeSigma
  • Novel 2-aryl-naphtho[1,2-d]oxazole derivatives as potential PTP-1B inhibitors showing antihyperglycemic activities.
Novel 2-aryl-naphtho[1,2-d]oxazole derivatives as potential PTP-1B inhibitors showing antihyperglycemic activities.
European journal of medicinal chemistry (2008-04-26)
Atul Kumar, Pervez Ahmad, Ram Awatar Maurya, A B Singh, Arvind K Srivastava
ABSTRACT
A series of 2-aryl-naphtho[1,2-d]oxazole derivatives have been synthesized and evaluated for PTP-1B inhibitory activity. The compounds have been screened in vivo for antidiabetic activity under sucrose loaded model (SLM), sucrose-challenged streptozotocin-induced diabetic rat model (STZ-S) and db/db mice model. Compounds 8 and 12 have shown promising PTP-1B inhibitory activity, significant antidiabetic activity, moderate lipid and triglyceride lowering activity.
Sigma-Aldrich
Glyburide, meets USP testing specifications