MilliporeSigma
  • Epstein-Barr Virus LMP1 Promotes Syntenin-1- and Hrs-Induced Extracellular Vesicle Formation for Its Own Secretion To Increase Cell Proliferation and Migration.
Epstein-Barr Virus LMP1 Promotes Syntenin-1- and Hrs-Induced Extracellular Vesicle Formation for Its Own Secretion To Increase Cell Proliferation and Migration.
mBio (2020-06-18)
Dingani Nkosi, Li Sun, Leanne C Duke, Nilkumar Patel, Sunil K Surapaneni, Mandip Singh, David G Meckes
ABSTRACT
Extracellular vesicles (EVs) are important mediators of cell-to-cell communication that are involved in both normal processes and pathological conditions. Latent membrane protein 1 (LMP1) is a major viral oncogene that is expressed in most Epstein-Barr virus (EBV)-associated cancers and secreted in EVs. LMP1-modified EVs have the ability to influence recipient cell growth, migration, and differentiation and regulate immune cell function. Despite the significance of LMP1-modified EVs in EBV malignancies, very little is understood about how this protein hijacks the host EV pathway for secretion. Using the biotin identification (BioID) method, we identified LMP1-proximal interacting proteins that are known to play roles in EV formation and protein trafficking. Analysis of the identified LMP1-interacting proteins revealed an enrichment in the ESCRT pathway and associated proteins, including CD63, Syntenin-1, Alix, TSG101, Hrs, and charged multivesicular body proteins (CHMPs). LMP1 transcriptionally upregulated and increased the protein expression of EV biogenesis and secretion genes. Nanoparticle tracking and immunoblot analysis revealed reduced levels of LMP1 EV packaging and of vesicle production following the knockdown of Syntenin-1, Alix, Hrs, and TSG101, with altered endolysosomal trafficking observed when Syntenin-1 and Hrs expression was reduced. Knockdown of specific ESCRT-III subunits (CHMP4B, -5, and -6) impaired LMP1 packaging and secretion into EVs. Finally, we demonstrate that the efficient secretion of LMP1-modified EVs promotes cell attachment, proliferation, and migration and tumor growth. Together, these results begin to shed light on how LMP1 exploits host ESCRT machinery to direct the incorporation of the viral oncoprotein into the EV pathway for secretion to alter the tumor microenvironment.IMPORTANCE LMP1 is a notable viral protein that contributes to the modification of EV content and tumor microenvironment remodeling. LMP1-modified EVs enhance tumor proliferation, migration, and invasion potential and promote radioresistance. Currently, the mechanisms surrounding LMP1 incorporation into the host EV pathways are not well understood. This study revealed that LMP1 utilizes Hrs, Syntenin-1, and specific components of the ESCRT-III complex for release from the cell, enhancement of EV production, and metastatic properties of cancer cells. These findings begin to unravel the mechanism of LMP1 EV trafficking and may provide new targets to control EBV-associated cancers.
Sigma-Aldrich
Doxycycline hydrochloride