MilliporeSigma
Hansch analysis of veratric acid derivatives as antimicrobial agents.
European journal of medicinal chemistry (2008-07-04)
Balasubramanian Narasimhan, Sucheta Ohlan, Ruchita Ohlan, Vikramjeet Judge, Rakesh Narang
ABSTRACT
The synthesis, characterization and antimicrobial evaluation of a new series of veratric acid derivatives are presented. Preliminary in vitro antimicrobial activity of the title compounds was assessed against a panel of microorganisms including Gram-positive and Gram-negative bacteria and fungi. Some of the veratric acid derivatives exhibited significant in vitro antimicrobial activity. QSAR investigation applied to find a correlation between different physicochemical parameters of the veratric acid derivatives and their antimicrobial activity indicated the importance of topological parameters in describing the antimicrobial activity.
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
3,4-Dimethoxybenzoic acid, ≥99%