MilliporeSigma
  • Vasoactive Intestinal Polypeptide (VIP) in the Intestinal Mucosal Nerve Fibers in Dogs with Inflammatory Bowel Disease.
Vasoactive Intestinal Polypeptide (VIP) in the Intestinal Mucosal Nerve Fibers in Dogs with Inflammatory Bowel Disease.
Animals : an open access journal from MDPI (2020-10-02)
Andrzej Rychlik, Sławomir Gonkowski, Jarosław Całka, Krystyna Makowska
ABSTRACT
Canine inflammatory bowel disease (IBD) is a group of enteropathies with nonspecific chronic symptoms and poorly understood etiology. Many aspects connected with IBD are not understood. One of them is the participation of the intestinal nervous system in the development of pathological processes. Thus, this study aimed to demonstrate changes in the density of intramucosal nerve fibers containing vasoactive intestinal polypeptide (VIP)-one of the most important intestinal nervous factors caused by the various stages of IBD development. Mucosal biopsy specimens collected from the duodenum, jejunum and descending colon of healthy dogs and dogs with varied severity of IBD were included in the experiment. The density of VIP-like immunoreactive (VIP-LI) nerves was determined by a single immunofluorescence technique and a semi-quantitative method consisting in VIP-LI fiber counts in the field of view (0.1 mm2). The obtained results indicate that IBD induces changes in the density of mucosal VIP-LI nerve fibers in the canine gastrointestinal tract. The initial decrease is followed by an increase in VIP-like immunoreactivity in successive stages of the disease. These observations show that VIP is a neuronal factor that participates in the pathological processes connected with canine IBD. The observed changes probably result from the neuroprotective and/or adaptive properties of VIP. Protective and adaptive reactions induced by inflammation aim to protect the GI tract against damage by proinflammatory factors and ensure the homeostasis in the enteric nervous system (ENS) under the conditions changed by the disease process.
Sigma-Aldrich
Vasoactive Intestinal Peptide human, porcine, rat, synthetic, ≥95% (HPLC), powder
Pricing and availability is not currently available.