MilliporeSigma
  • Prognostic impact of somatostatin receptor expression in advanced bladder cancer.
Prognostic impact of somatostatin receptor expression in advanced bladder cancer.
Urologic oncology (2020-08-11)
Moritz Maas, Lisa Mayer, Jörg Hennenlotter, Viktoria Stühler, Simon Walz, Marcus Scharpf, Ursula Kühs, Tim Neumann, Arnulf Stenzl, Tilman Todenhöfer
ABSTRACT
Somatostatin receptors (SSTR) recently have been identified as potential targets for treatment of solid tumors. Furthermore, they have been shown to be of high relevance for tumor biology and prognosis in various types of cancer. However, there is a lack of clinical data for SSTR in bladder cancer (BC). Aim of this study was to determine the expression of all relevant somatostatin receptor subtypes in benign urothelium and tumor tissue of patients with muscle invasive BC. Furthermore, their potential role as prognostic factor for cancer-specific survival (CSS) and overall survival (OS) was evaluated. The collective included BC and benign urothelium tissue of 103 patients (Median age 69; range 32-84, 79 male, 24 female) who underwent a radical cystectomy. A tissue microarray with subsequent immunohistochemical staining was used to assess membranous expression of SSTR1-5. Results were correlated to clinical and histopathological data as well as CSS and OS. Expressions of SSTR1-4 were significantly decreased in BC compared to benign urothelium (P < 0.002 each), whereas SSTR5 expression was increased (P = 0.0017). Expression of SSTR1 was associated with organ-confined disease (≤pT2) (P = 0.0477). No correlation between SSTR1-5 expression and N- and M-stage was observed. Univariate analyses showed a significantly longer CSS and OS in patients with high expression of SSTR3 (P = 0.0316 and 0.0044). Multivariate analyses confirmed SSTR3 expression as independent marker of improved CSS and OS (P = 0.0324 and 0.0076). The majority of somatostatin receptor subtypes exhibit decreased expression in BC compared to benign bladder tissue. Expression of SSTR3 is an indicator for favorable prognosis in patients with muscle-invasive BC. These results support preclinical investigations using somatostatin receptor analogues such as octreotide to influence BC growth.