MilliporeSigma
Semisynthesis and biological activity of stemofoline alkaloids.
Journal of natural products (2009-02-19)
Morwenna C Baird, Stephen G Pyne, Alison T Ung, Wilford Lie, Thanapat Sastraruji, Araya Jatisatienr, Chaiwat Jatisatienr, Srisulak Dheeranupattana, Jaturong Lowlam, Sukanya Boonchalermkit
ABSTRACT
The semisynthesis of the Stemona alkaloids (3'R)-stemofolenol (1), (3'S)-stemofolenol (2), methylstemofoline (3), and (3'S)-hydroxystemofoline (5) and the unnatural analogues (11E)-methylstemofoline (15) and 3'R-hydroxystemofoline (11) has been achieved starting from (11Z)-1',2'-didehydrostemofoline (4). This synthesis allowed, for the first time, access to diastereomerically enriched samples of 1 and 2 and the assignment of their absolute configurations at C-3'. These compounds were obtained in sufficient quantities to allow for their biological testing. In a quantitative assay as AChE inhibitors, (11Z)-1',2'-didehydrostemofoline (4) and (3'S)-hydroxystemofoline (5) were found to be the most active.