MilliporeSigma
  • Piperidine-4-carboxamide as a new scaffold for designing secretory glutaminyl cyclase inhibitors.
Piperidine-4-carboxamide as a new scaffold for designing secretory glutaminyl cyclase inhibitors.
International journal of biological macromolecules (2020-12-30)
K V Dileep, Naoki Sakai, Kentaro Ihara, Miyuki Kato-Murayama, Akiko Nakata, Akihiro Ito, D M Sivaraman, Jay W Shin, Minoru Yoshida, Mikako Shirouzu, Kam Y J Zhang
ABSTRACT
Alzheimer's disease (AD), a common chronic neurodegenerative disease, has become a major public health concern. Despite years of research, therapeutics for AD are limited. Overexpression of secretory glutaminyl cyclase (sQC) in AD brain leads to the formation of a highly neurotoxic pyroglutamate variant of amyloid beta, pGlu-Aβ, which acts as a potential seed for the aggregation of full length Aβ. Preventing the formation of pGlu-Aβ through inhibition of sQC has become an attractive disease-modifying therapy in AD. In this current study, through a pharmacophore assisted high throughput virtual screening, we report a novel sQC inhibitor (Cpd-41) with a piperidine-4-carboxamide moiety (IC50 = 34 μM). Systematic molecular docking, MD simulations and X-ray crystallographic analysis provided atomistic details of the binding of Cpd-41 in the active site of sQC. The unique mode of binding and moderate toxicity of Cpd-41 make this molecule an attractive candidate for designing high affinity sQC inhibitors.
Sigma-Aldrich
1-Benzylimidazole, 99%