MilliporeSigma
  • Effect of essential oils, such as raspberry ketone and its derivatives, on antiandrogenic activity based on in vitro reporter gene assay.
Effect of essential oils, such as raspberry ketone and its derivatives, on antiandrogenic activity based on in vitro reporter gene assay.
Bioorganic & medicinal chemistry letters (2010-03-17)
Yoshihisa Ogawa, Miki Akamatsu, Yudai Hotta, Akifumi Hosoda, Hiroto Tamura
ABSTRACT
The effect of essential oils, such as raspberry ketone, on androgen (AR) receptor was investigated using a MDA-kb2 human breast cancer cell line for predicting potential AR activity. Among them, eugenol had the highest AR antagonistic activity with its IC(50) value of 19 microM. Raspberry ketone, which has threefold higher anti-obese activity than that of capsaicin, also had AR antagonist activity with its IC(50) value of 252 microM. Based on these findings, a more precise CoMFA model was proposed as follows: pIC(50) [log (1/IC(50))]=3.77+[CoMFA field terms] (n=39, s=0.249, r(2)=0.834, s(cv)=0.507, q(2)=0.311 (three components).
Sigma-Aldrich
Capsaicin, from Capsicum sp., ≥50% (HPLC)
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Supelco
Eugenol, PESTANAL®, analytical standard
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Sigma-Aldrich
4-tert-Octylphenol, 97%
Sigma-Aldrich
4-(4-Hydroxyphenyl)-2-butanone, ≥98%, FCC, FG
Sigma-Aldrich
4-(4-Hydroxyphenyl)-2-butanone, natural, ≥98%, FCC, FG
Sigma-Aldrich
4-(4-Hydroxyphenyl)-2-butanone, 99%
Sigma-Aldrich
Capsaicin, ≥95%, from Capsicum sp.
Sigma-Aldrich
Capsaicin, natural
Supelco
4-tert-Octylphenol, analytical standard