MilliporeSigma
Benzyl prolinate derivatives as novel selective KCC2 blockers.
Bioorganic & medicinal chemistry letters (2010-03-20)
Cécile Pégurier, Nathalie Bosman, Philippe Collart, Marie-Laure Delporte, Karine Leclercq, Sébastien Lengelé, Ananda Kumar Kanduluru, Stéphane Meunier, Nathalie Pacico, Lakshmana Rao Vadali, Alain Wagner, Christian Wolff, Laurent Provins
ABSTRACT
The discovery and optimization of a novel class of selective submicromolar KCC2 blockers is described. Details of synthesis and SAR are given together with ADME properties of selected compounds. A methylsulfone residue on the R(1) phenyl group improved the overall general profile of these prolinate derivatives.
Sigma-Aldrich
Furosemide